Buffer, TBS solution 10X concentrate (TRIS buffered saline)
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
R029 R030
R029EA 113 EUR
R029 R030
Buffer, TBS solution 10X concentrate (TRIS buffered saline)
Buffers Buffers voor elektroforese Wasbuffers
The TBS solution is primarily used as a wash and blocking buffer diluent for Western blot, ELISA and immunoassays.
10X TBS: Tris buffered saline [10X]: 100mM Tris HCl, 1.5M NaCl, pH 7.5