Buffer, TE Solution (TRIS-EDTA buffer) pH 8,0, Low EDTA for Molecular Biology
Leverancier: PanReac AppliChem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A8569.0500 A8569.1000
A8569.0500EA 27.2 EUR
A8569.0500 A8569.1000
Buffer, TE Solution (TRIS-EDTA buffer) pH 8,0, Low EDTA for Molecular Biology
Buffers Kloonbuffers
  • DNases/RNases/Proteases: not detectable
  • pH (20°C; adjusted with HCl): 8,0 ± 0,1
Composition: EDTA · Na2: 0,1 mM and Tris: 10 mM.