Oxytetracycline hydrochloride
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31357.01
31357.02
SERA31357.01EA
0
EUR
SERA31357.01
SERA31357.02
Oxytetracycline hydrochloride
Oxytetracycline hydrochloride
Formule:
C₂₂H₂₅ClN₂O₉ MW: 496.90 g/mol |
MDL Nummer:
MFCD00135815 CAS nummer: 2058-46-0 EINECS: 218-161-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...