Dialysis tubing Spectra/Por® 5
Leverancier: Spectrum Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
132754 132757 132757T
734-0697EA 313 EUR
734-0697 734-0698 734-2647
Dialysis tubing Spectra/Por® 5
Slangen Dialyseslangen
MWCO 12 - 14 kD, regenerated cellulose
  • Transparent and flexible
Supplied dry, preservative: glycerol
Order Now
Specificaties
Environmentally Preferable