Vials, Vacule®, WHEATON®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
651905 651907 651954
WHEA651905EA 440 EUR
WHEA651905 WHEA651907 WHEA651954
Vials, Vacule®, WHEATON®
flesjes
These vacule vials made of 33 low extractable clear borosilicate glass feature a heavy wall construction designed for lyophilisation and freeze-drying applications.
  • Vials have controlled ID, for accurate stopper fit
  • Screw threads eliminate use of crimp seal
Certificaten: Meets USP Type I and ASTM E 438 Type I.
Bestelinformatie: Closures for 4 ml vials have to be ordred separately.
Leveringsinformatie: Vials, 5 and 10 ml are supplied with unlined black phenolic cap.