Sluitkegels, Combi-Stoppers
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4495101 4495152 4495209
612-0100EA 52.8 EUR
612-0100 BRAU4495152 BRAU4495209
Sluitkegels, Combi-Stoppers
Stoppen PE/LDPE/HDPE stoppen
These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
  • Unique multifunctional design
  • Sterile closing of pre-filled syringes and all kinds of open IV-accesses in IV-Sets
Available in red, blue and white.