Accessories for centurion helmets, Connect®
Leverancier: CENTURION SAFETY PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S57CE S598 S570
111-0898EA 39.43 EUR
111-0898 111-0903 111-1524
Accessories for centurion helmets, Connect®
Hoofdbescherming Veiligheidshelmen
Hinged and individually adjustable face shield attachment of polyetherimide (PEI). For use in hot factories.
  • Combine with: Connect ® clear shield high temp S598 (SKU: 0101028) Connect ® shield tinted High Temp S603 (SKU: 0101029)
  • Fits Vulcan Helmet
Weight: 150 g
EN 166 (depending on the face plate)