Bump cap, Cool Cap
Leverancier: CENTURION SAFETY PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S28RB S28G S28K S28R S28NB
CENUS28RBEA 53.2 EUR
CENUS28RB CENUS28G CENUS28K CENUS28R CENUS28NB
Bump cap, Cool Cap
Hoofdbescherming Stootpetten
This brushed cotton bump cap is fitted over an ABS liner to provide lightweight head protection where high impact resistance is not essential, but where there is a risk of minor bumps or scrapes to the head.
  • Shell made of lightweight ABS
  • Breathable thanks to sleek three panel design with improved side ventilation
  • Removable foam side inserts provide 45% more shock absorbency than standard ABS liner
Suitable applications include automotive, airlines, transportation, shipping, railways and engineering.
For head sizes 54 to 59 cm
Peak length: 50 mm
Weight: 155 g
Certificaten: EN 812