Flame-retardant overalls with bib and braces, FLAM Bib'N'Brace
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100328540C46 100328540C48 100328540C50 100328540C52 100328540C54 100328540C56 100328540C58 100328540C60 100328540C62
KWIN100328540C46EA 183.9 EUR
KWIN100328540C46 KWIN100328540C48 KWIN100328540C50 KWIN100328540C52 KWIN100328540C54 KWIN100328540C56 KWIN100328540C58 KWIN100328540C60 KWIN100328540C62
Flame-retardant overalls with bib and braces, FLAM Bib'N'Brace
Werkkleding
Trousers made of a cotton/polyester fabric with a special treatment, providing protection against heat and open flames. The trousers also feature fluorescent fabrics for an improved visibility.
  • Two-way front zip
  • Two loose-hanging, detachable chest pockets, one with flap and concealed snap fasteners, one with extra pocket and pen pocket
  • Button and loop for sheath knife
  • Two front pockets, two loose-hanging pockets with extra width, one with extra pocket, one with three tool pockets and tool loops, hammer loop
  • Two back pockets with extra width, leg pocket with phone pocket and tool pocket
  • Folding rule pocket over side seam, with tool pocket and button and loop for sheath knife
  • Rrounded knees with knee pockets
Ideal for applications that require protection against flames, open arcs and welding sparks.
Weight: 365 g/m²
Certificaten: EN 61482-1-2 (class 1); EN ISO 11612 (A1, B1, C1)