Rain trousers, Bangkok 6360
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6360/B90/S 6360/B90/M 6360/B90/L 6360/B90/1XL 6360/B90/2XL 6360/B90/3XL 6360/Y58/M 6360/Y58/L 6360/Y58/1XL 6360/Y58/S 6360/A41/S 6360/Y58/2XL 6360/A41/M 6360/A41/L 6360/A41/1XL 6360/A41/2XL 6360/Y58/3XL 6360/A41/3XL
113-5962EA 29.5 EUR
113-5962 113-5961 113-5960 113-5969 113-5970 113-5971 113-5964 113-5963 113-5972 113-5965 113-5959 113-5973 113-5958 113-5957 113-5966 113-5967 113-5974 113-5968
Rain trousers, Bangkok 6360
Werkkleding
Waterproof and windproof trousers made of Flexothane® Essential: 100% polyester knitting with PU coating.
  • Supple, stretchable material: Comfortable to wear and noiseless
  • High frequency welded seams
  • Legs adjustable by press studs
  • Elasticated waist
Weight: 170 g/m²
Certificaten: ENV 343 (3-1)