Winter jacket, Treban 193A
Leverancier: Sioen
Ratings:
Avg. Ratings: 3.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 3.0 out of 5
Was this review helpful? Yes No
Yes: 0 No: 0 
Personnel / COURCELLES LES MONTBÉLIARD
Yes, I recommend
Bonjour , belle veste chaude , mais un peu lourde !
Sioen
113-7013EA 80.2 EUR
113-7013 113-7012 113-7011 113-7010 113-7009 113-7008
Winter jacket, Treban 193A
Werkkleding
Rain and cold resistant jacket made of 100% polyester fabric with PVC coating. Upper body in fleece lining, lower body in quilt lining. With reflective striping on sleeves, chest and back and microprism insert on front flap.
  • Straight collar with foldaway hood
  • Zip closure under flap with touch and close fastening
  • Two inset chest pockets and two patched pockets, phone pocket, sleeve pocket and pen pocket, loop for badge
  • Round inset sleeves, elastic at end with narrowing by touch and close fastening
  • Elasticated waist in the back
Back length: 85 cm
Weight: 330 g/m²
Certificaten: EN 343, EN 14058 (3XXXX)