Shorts, Originals 275 FAS
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100288940C44 100288940C46 100288940C48 100288940C50 100288940C52 100288940C54 100288940C56 100288940C58 100288940C60 100288940C62 100288541C44 100288541C46 100288541C48 100288541C50 100288541C52 100288541C54 100288541C56 100288541C58 100288541C60 100288541C62
KWIN100288940C44EA 61.9 EUR
KWIN100288940C44 KWIN100288940C46 KWIN100288940C48 KWIN100288940C50 KWIN100288940C52 KWIN100288940C54 KWIN100288940C56 KWIN100288940C58 KWIN100288940C60 KWIN100288940C62 KWIN100288541C44 KWIN100288541C46 KWIN100288541C48 KWIN100288541C50 KWIN100288541C52 KWIN100288541C54 KWIN100288541C56 KWIN100288541C58 KWIN100288541C60 KWIN100288541C62
Shorts, Originals 275 FAS
Werkkleding
Short working trousers made of FAS®, Fristads®/Kansas® best twill made of 100% cotton.
  • Two tuckable, loose-hanging pockets, one with extra pocket, one with three small pockets and tool pocket
  • Two front and back pockets
  • Folding ruler pocket, mobile phone pocket, pen pocket, tool pocket
  • Button and loop for sheath knife, hammer loop, D-ring
  • Öko-Tex® certified
Weight: 375 g/m²