Trousers for floor layers and carpet fitters, Fristads® AD-265K
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100083930C148 100083930C50
KWIN100083930C148EA 0 EUR
KWIN100083930C148 KWIN100083930C50
Trousers for floor layers and carpet fitters, Fristads® AD-265K
Werkkleding
Made from Fristads Polydex®, 100% polyester; with napped inner. Can be combined: with knee protection, KS-950 knee protection (thickness: 25 mm), KT-952 knee protection (thickness: 13 mm).
  • Two loose-hanging, detachable, front, nail pockets – one with an extra compartment, the others with 3 smaller compartments and tool loops
  • Two particularly roomy front and back pockets; folding-rule pocket; pencil pocket; tool pocket; phone pocket; leg pocket with flap
  • Two hammer loops, one of them adjustable
  • Knife button; knee pockets (Cordura® reinforced)
  • Additional floor layer pocket
Weight: 245 g/m²