Trousers, Fristads® AD-255K
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100082940C146 100082940C148 100082940C150 100082940C152 100082940C154 100082940C156 100082940C44 100082940C46 100082940C48 100082940C50 100082940C52 100082940C54 100082940C56 100082940C58 100082940C60 100082940D100 100082940D104 100082940D108 100082940D112 100082940D116 100082940D96
KWIN100082940C146EA 87.9 EUR
KWIN100082940C146 KWIN100082940C148 KWIN100082940C150 KWIN100082940C152 KWIN100082940C154 KWIN100082940C156 KWIN100082940C44 KWIN100082940C46 KWIN100082940C48 KWIN100082940C50 KWIN100082940C52 KWIN100082940C54 KWIN100082940C56 KWIN100082940C58 KWIN100082940C60 KWIN100082940D100 KWIN100082940D104 KWIN100082940D108 KWIN100082940D112 KWIN100082940D116 KWIN100082940D96
Trousers, Fristads® AD-255K
Werkkleding
Work trousers made from Polydex®, 100% polyester, with napped inner.
  • Material is reistant to dirt, oil and water, particularly easy to care for
  • Two loose-hanging, detachable, front, nail pockets – one with an extra compartment, the others with 3 smaller compartments and tool loops
  • Two particularly roomy front and back pockets
  • Folding-rule pocket; pencil pocket; tool pocket; phone pocket
  • Leg pocket with flap, Cordura® knee pockets
  • Two hammer loops, one of them adjustable; knife button
Material weight: 245 g/m²
Bestelinformatie: Can be combined: with knee protection, KS-950 knee protection (thickness: 25mm), KT-952 knee protection (thickness: 13mm).