Trousers, ladies, Fristads® BM-259
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10012190034 10012190036 10012190038 10012190040 10012190042 10012190044 10012190046 10012190048
KWIN10012190034EA 80.9 EUR
KWIN10012190034 KWIN10012190036 KWIN10012190038 KWIN10012190040 KWIN10012190042 KWIN10012190044 KWIN10012190046 KWIN10012190048
Trousers, ladies, Fristads® BM-259
Werkkleding
Ladies trousers made from 100% cotton twill. Knee and material pockets reinforced with Cordura®.
  • Two loose-hanging, detachable material pockets, with extra pocket and three small pockets
  • Two roomy front and back pockets; hammer loop; D-ring
  • Folding-rule pocket, comfortably placed on the side seam, with tool holder and pen pocket
  • Telephone pocket with flap
  • Loop and button for bladed tools
  • Leg pocket with flap, one extra pocket with concealed ID card holder
  • Preformed knee with knee pockets; internal opening; knee protection to be positioned at various heights
Weight: 300 g/m²