Industrial jacket, Fristads® PR54-420, design A, black
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
847940730XL 847940730XS 8479409402XL 847940940L 847940940M 847940940S 847940940XL 847940940XS 8479409412XL 847940941L 847940941M 847940941S 847940941XL 847940941XS 8479401312XL 847940131L 847940131M 847940131S 847940131XL 847940131XS 8479402102XL 847940210L 847940210M 847940210S 847940210XL 847940210XS 8479402312XL 847940231L 847940231M 847940231S 847940231XL 847940231XS 8479403312XL 847940331XL 8479404402XL 847940440L 847940440M 847940440S 847940440XL 847940440XS 8479405202XL 847940520L 847940520M 847940520S 847940520XL 847940520XS 8479405302XL 847940530L 847940530M 847940530S 847940530XL 847940530XS 8479405392XL 847940539L 847940539M 847940539S 847940539XL 847940539XS 8479406402XL 847940640L 847940640M 847940640S 847940640XL 847940640XS 8479407302XL 847940730L 847940730M 847940730S
113-5038EA 0 EUR
113-5038 113-03245 113-5053 113-03269 113-03270 113-03271 113-5052 113-03272 113-5043 113-03249 113-03250 113-03251 113-5042 113-03252 113-5045 113-03253 113-03254 113-03255 113-5044 113-03256 113-5041 113-03246 113-5394 113-03247 113-5040 113-03248 113-5047 113-03257 113-03258 113-03259 113-5046 113-03260 113-5049 113-5048 113-5051 113-03265 113-03262 113-03263 113-5050 113-03264 113-5028 113-03226 113-03227 113-03228 113-5027 113-03229 113-5034 113-03234 113-03235 113-03236 113-5033 113-03237 113-5032 113-03230 113-03231 113-03232 113-5029 113-03233 113-5037 113-03238 113-03239 113-03240 113-5036 113-03241 113-5039 113-03242 113-03243 113-03244
Industrial jacket, Fristads® PR54-420, design A, black
Werkkleding
Thanks to its durability and high degree of comfort, the PR54 jacket is perfectly suited to workshops and industrial and service enterprises. The material is made from 65% polyester and 35% cotton. The inside is brushed and as a result conveys a pleasantly comfortably cotton feeling. The outside is also resistant to the roughest demands and will not fade even after countless washes.
  • Zip fastening and plastic press studs
  • Two breast pockets with flap, two front pockets
  • Sleeve pocket, mobile phone pocket, pen pocket, inner pocket
  • Back pleats, adjustable waist
  • 12 decorative colours available
Weight: 260 g/m²
Basic colour: Black