Industrial jacket, Fristads® PR54-420, design B, black
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8489402102XL 848940210L 848940210M 848940210S 848940210XL 848940210XS 8489402312XL 848940231L 848940231M 848940231S 848940231XL 848940231XS 8489403312XL 848940331L 848940331M 848940331S 848940331XL 848940331XS 8489404402XL 848940440L 848940440M 848940440S 848940440XL 848940440XS 8489405202XL 848940520L 848940520M 848940520S 848940520XL 848940520XS 8489405302XL 848940530L 848940530M 848940530S 848940530XL 848940530XS 8489405392XL 848940539L 848940539M 848940539S 848940539XL 848940539XS 8489406402XL 848940640L 848940640M 848940640S 848940640XL 848940640XS 8489407302XL 848940730L 848940730M 848940730S 848940730XL 848940730XS 8489409402XL 848940940L 848940940M 848940940S 848940940XL 848940940XS 8489409412XL 848940941L 848940941M 848940941S 848940941XL 848940941XS
113-5138EA 0 EUR
113-5138 113-03437 113-03438 113-03439 113-5137 113-03440 113-5144 113-03449 113-03450 113-03451 113-5143 113-03452 113-5146 113-03453 113-03454 113-03455 113-5145 113-03456 113-5148 113-03457 113-03458 113-03459 113-5147 113-03460 113-5128 113-03417 113-03418 113-03419 113-5127 113-03420 113-5132 113-03425 113-03426 113-03427 113-5131 113-03428 113-5130 113-03421 113-03422 113-03423 113-5129 113-03424 113-5134 113-03429 113-03430 113-03431 113-5133 113-03432 113-5136 113-03433 113-03434 113-03435 113-5135 113-03436 113-5150 113-03461 113-03462 113-03463 113-5149 113-03464 113-5140 113-03441 113-03442 113-03443 113-5139 113-03444
Industrial jacket, Fristads® PR54-420, design B, black
Werkkleding
Thanks to its durability and high degree of comfort, the PR54 jacket is perfectly suited to workshops and industrial and service enterprises. The material is made from 65% polyester and 35% cotton. The inside is brushed and as a result conveys a pleasantly comfortably cotton feeling. The outside is also resistant to the roughest demands and will not fade even after countless washes.
  • Zip fastening and plastic press studs
  • Two breast pockets with flap, two front pockets
  • Sleeve pocket, mobile phone pocket, pen pocket, inner pocket
  • Back pleats, adjustable waist
More Product Information
Order Now
Specificaties
Model PR54-420, design B, black
Environmentally Preferable
Kledingstuk toepassing Services,Industrial
Risk area
Geslacht Voor mannen
Sleeve or leg length Long
Sluitingstype Met ritssluiting
Stof 65% polyester, 35% cotton
Gewicht 260 g/m²