Trousers, Fristads® PR54-220, Design B, Navy blue
Leverancier: Kwintet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Fristads®
113-01144EA 0 EUR
113-01144 113-01145 113-01142 113-01143 113-01140 113-01141 113-01148 113-01149 113-01146 113-01147 113-01155 113-01156 113-01153 113-01154 113-01151 113-01152 113-01150 113-02802 113-02803 113-02800 113-02801 113-01159 113-01157 113-01158 113-01164 113-01165 113-01160 113-01161 113-02813 113-02814 113-02811 113-02812 113-02810 113-02808 113-02809 113-02806 113-02807 113-02804 113-02805 113-01171 113-01172 113-02824 113-02825 113-02822 113-02823 113-02820 113-02821 113-02819 113-02817 113-02818 113-02815 113-02816 113-01100 113-01067 113-02795 113-01068 113-01101 113-01066 113-02835 113-01108 113-01109 113-02836 113-01106 113-02833 113-01107 113-02834 113-01104 113-02831 113-02798 113-01105 113-02832 113-02799 113-01069 113-01102 113-02796 113-02797 113-01103 113-02830 113-02828 113-02829 113-02826 113-02827 113-01078 113-01111 113-01112 113-01076 113-01077 113-01110 113-01074 113-01075 113-01072 113-01073 113-02846 113-01119 113-02847 113-01117 113-02844 113-01118 113-02845 113-01115 113-02842 113-02843 113-01116 113-02840 113-01113 113-02841 113-01114 113-02839 113-02837 113-02838 113-01070 113-01071 113-01122 113-01089 113-01123 113-02850 113-01087 113-01120 113-01088 113-01121 113-01085 113-01086 113-01083 113-01084 113-01128 113-01129 113-02853 113-01126 113-01127 113-02854 113-02851 113-01124 113-01125 113-02852 113-02848 113-02849 113-01081 113-01082 113-01080 113-01133 113-01134 113-01131 113-01098 113-01132 113-01099 113-01096 113-01097 113-01130 113-01094 113-01095 113-01139 113-01137 113-01138 113-01135 113-01136 113-01092 113-01093 113-01090 113-01091
Trousers, Fristads® PR54-220, Design B, Navy blue
Werkkleding
Thanks to their durability and comfort, these trousers are perfectly suitable for use in garages as well as in industrial and service companies. The material is made of 65% polyester and 35% cotton. The inside is brushed and provides a comfortable cotton feel. The outside is very hardwearing and does not bleach, even after countless washes.
  • Two front and back pockets
  • Two leg pockets - one with flap, telephone pocket, pen pocket, tool bags
  • Adjustable waist, plastic snap buttons
  • Colour accents on pocket edges and leg pocket flap
Weight: 260 g/m²
Basic colour: navy blue