Gilets, Fristads® PR54-521 Design A, dark grey
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8519411312XL 851941131L 851941131M 851941131S 851941131XL 851941131XS 8519412102XL 851941210L 851941210M 851941210S 851941210XL 851941210XS 8519412312XL 851941231L 851941231M 851941231S 851941231XL 851941231XS 8519413312XL 851941331L 851941331M 851941331S 851941331XL 851941331XS 8519414402XL 851941440L 851941440M 851941440S 851941440XL 851941440XS 8519415202XL 851941520L 851941520M 851941520S 851941520XL 851941520XS 8519415302XL 851941530L 851941530M 851941530S 851941530XL 851941530XS 8519415392XL 851941539L 851941539M 851941539S 851941539XL 851941539XS 8519416402XL 851941640L 851941640M 851941640S 851941640XL 851941640XS 8519417302XL 851941730L 851941730M 851941730S 851941730XL 851941730XS 8519419402XL 851941940L 851941940M 851941940S 851941940XL 851941940XS 8519419412XL 851941941L 851941941M 851941941S 851941941XL 851941941XS
113-5310EA 0 EUR
113-5310 113-03807 113-03808 113-03809 113-5309 113-03810 113-5306 113-03799 113-03800 113-03801 113-5305 113-03802 113-5312 113-03811 113-03812 113-03813 113-5311 113-03814 113-5314 113-03815 113-03816 113-03817 113-5313 113-03818 113-5316 113-03819 113-03820 113-03821 113-5315 113-03822 113-5296 113-03779 113-03780 113-03781 113-5295 113-03782 113-5300 113-03787 113-03788 113-03789 113-5299 113-03790 113-5298 113-03783 113-03784 113-03785 113-5297 113-03786 113-5302 113-03791 113-03792 113-03793 113-5301 113-03794 113-5304 113-03795 113-03796 113-03797 113-5303 113-03798 113-5318 113-03823 113-03824 113-03825 113-5317 113-03826 113-5308 113-03803 113-03804 113-03805 113-5307 113-03806
Gilets, Fristads® PR54-521 Design A, dark grey
Werkkleding
Vest made from 65% polyester and 35% cotton; napped inner.
  • Zip fastening; 2 breast pockets with flap; front pockets; inner pocket
  • Back pleats, waistband adjustable using press studs, plastic press studs
  • Colour contrasting: Upper collar, bottom edge of zip cover, pocket edges and section on front and back
Material weight: 260 g/m²
Basic colour: Dark grey
Order Now
Specificaties
Model PR54-521, Design B
Environmentally Preferable
Kledingstuk toepassing General Purpose
Risk area
Geslacht Voor mannen
Sleeve or leg length Short
Sluitingstype Met ritssluiting
Stof 65% polyester and 35% cotton