Safety shoes, lace-up, Ireland
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0082183/38 0082183/39 0082183/48 0082183/35 0082183/36 0082183/37
114-2384EA 80.6 EUR
114-2384 114-2385 114-2394 114-2381 114-2382 114-2383
Safety shoes, lace-up, Ireland
Schoenen en laarzen
Safety shoe with Air-Light® - nylon lining, dries quickly and stays pleasantly fresh. Air-Relax® insole made of micro-perforated carbon.
  • Upper material: 2 mm thick full grain leather
  • Outsole: non-slip and abrasion-resistant 2-layer polyurethane (PU) sole
  • Safety cap: steel cap
  • Penetration resistance: steel midsole
Width: 10,5
Certificaten: ISO EN 20345, S3 SRC