Men's fleece jacket, Acode® Basecamp
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100171331M 100171331XL
KWIN100171331MEA 0 EUR
KWIN100171331M KWIN100171331XL
Men's fleece jacket, Acode® Basecamp
Werkkleding
Outdoor jacket made from 100% polyester fleece, with anti-pilling treatment.
  • Two side pockets, one sleeve pocket - all with zip closure
  • Hem with elasticated cord
Weight: 240 g/m²