Safety shoes, lace-up, Jalscand
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J0248/39 J0248/40 J0248/41 J0248/42 J0248/43 J0248/44 J0248/45 J0248/46 J0248/47
JALCJ0248/39EA 134 EUR
JALCJ0248/39 JALCJ0248/40 JALCJ0248/41 JALCJ0248/42 JALCJ0248/43 JALCJ0248/44 JALCJ0248/45 JALCJ0248/46 JALCJ0248/47
Safety shoes, lace-up, Jalscand
Schoenen en laarzen
Safety shoes made from hydrophobic full-grain leather with tongue made from leather, 3D lining and PU/nitrile rubber outer sole (heat resistant to 300 °C).
  • Multi-layer lining is permeated with microchannels and ensures optimal breathability
  • Anatomically shaped Confort-Tech insole; anti-static and antibacterial, with shock absorber heel
  • Softane™ dual-density PU provides excellent absorption of heel shocks and vibrations
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: black
Colour: Black
Certificaten: EN ISO 20345:2007; S3, HRO, SRC