Shirt, long sleeves, Legacy
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1007319402XL 1007319403XL 100731940L 100731940M 100731940S 100731940XL
KWIN1007319402XLEA 44.9 EUR
KWIN1007319402XL KWIN1007319403XL KWIN100731940L KWIN100731940M KWIN100731940S KWIN100731940XL
Shirt, long sleeves, Legacy
Werkkleding
Men's shirt in Luxe fabric made of 65% polyester and 35% cotton.
  • Pretreated against shrinkage
  • Yoke and action back pleat
  • Two chest pockets, pen pocket, mobile phone pocket
Weight: 177 g/m²