Thermo waistcoat, Match
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1007115402XL 1007115403XL 1007115404XL 100711540L 100711540M 100711540S 100711540XL 100711540XS
KWIN1007115402XLEA 54.9 EUR
KWIN1007115402XL KWIN1007115403XL KWIN1007115404XL KWIN100711540L KWIN100711540M KWIN100711540S KWIN100711540XL KWIN100711540XS
Thermo waistcoat, Match
Werkkleding
Insulating waistcoat for moderately cold environments. The waistcoat is manufactured from 100% polyester.
  • With plastic zip closure
  • Kidney protector
  • Elasticated back
  • Chest pocket with zip closure
Weight: 180 g/m²