Safety ankle boots, lace-up, Jaltarmac
Leverancier: Jallatte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J0250/40 J0250/41 J0250/42 J0250/43 J0250/44 J0250/45 J0250/46 J0250/47 J0250/39
JALCJ0250/40EA 151 EUR
JALCJ0250/40 JALCJ0250/41 JALCJ0250/42 JALCJ0250/43 JALCJ0250/44 JALCJ0250/45 JALCJ0250/46 JALCJ0250/47 JALCJ0250/39
Safety ankle boots, lace-up, Jaltarmac
Schoenen en laarzen
Safety boots specifically for road construction. Made from hydrophobic nubuck leather with heat resistant Softane™/NBR (nitrile rubber) outer sole (to 300 °C).
  • Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
  • Isolating multi-layer complex insole prevents from heat shocks which may occur when working on asphalt or floors with temperatures of 160 °C
  • Sole with lug tread for best anti-slip properties and improved shock absorption
  • Protective steel toecap (200 J)
Colour: Black/red
Certificaten: EN 345-1; S3, HI, HRO