Ladies polo shirt, Acode® Advanced, Cool Dry
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1002169402XL 100216940L 100216940M 100216940S 100216940XL
KWIN1002169402XLEA 27.8 EUR
KWIN1002169402XL KWIN100216940L KWIN100216940M KWIN100216940S KWIN100216940XL
Ladies polo shirt, Acode® Advanced, Cool Dry
Werkkleding
Functional polo shirt made of Cool Dry fabric piqué (100% polyester). The soft and flexible fabric transports moisture (sweat) away from the body. Ideal for outdoor activities and sports.
  • Fitted design
  • With neckline
  • Raglan sleeves
  • Collar made of piqué fabric
Weight: 150 g/m²