Belts, 992 SK
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100583540 100583940
113-6633EA 15.5 EUR
113-6633 113-6634
Belts, 992 SK
Werkkleding
Belt made of 100% polyamide
  • Antique brass or antique silver slide buckle
  • Width 40 mm