Hand cleansing pastes, Super Plum
Leverancier: Plum Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1043 1018 1015
PLUM1043EA 165 EUR
PLUM1043 115-1369 115-1368
Hand cleansing pastes, Super Plum
Huidverzorging Zepen en huidreinigingsmiddelen
Mild hand cleansing paste based on vegetable oils. Effectively but gently removes oil, oily dirt, printing inks and other ordinary dirt.
  • Without solvents
  • To be applied on wet hands
  • For dispenser systems MP2000
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904