Sweatshirt, Match
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1007823312XL 1007823313XL 1007823314XL 100782331L 100782331M 100782331S 100782331XL 100782331XS 1007825302XL 1007825303XL 1007825304XL 100782530L 100782530M 100782530S 100782530XL 100782530XS 1007825402XL 1007825403XL 1007825404XL 100782540L 100782540M 100782540S 100782540XL 100782540XS 1007827302XL 1007827303XL 1007827304XL 100782730L 100782730M 100782730S 100782730XL 100782730XS 100782930L 100782930M 100782930S 100782930XS 1007829302XL 1007829303XL 1007829304XL 100782930XL 1007829402XL 1007829403XL 1007829404XL 100782940L 100782940M 100782940S 100782940XL 100782940XS
KWIN1007823312XLEA 33.5 EUR
KWIN1007823312XL KWIN1007823313XL KWIN1007823314XL KWIN100782331L KWIN100782331M KWIN100782331S KWIN100782331XL KWIN100782331XS KWIN1007825302XL KWIN1007825303XL KWIN1007825304XL KWIN100782530L KWIN100782530M KWIN100782530S KWIN100782530XL KWIN100782530XS KWIN1007825402XL KWIN1007825403XL KWIN1007825404XL KWIN100782540L KWIN100782540M KWIN100782540S KWIN100782540XL KWIN100782540XS KWIN1007827302XL KWIN1007827303XL KWIN1007827304XL KWIN100782730L KWIN100782730M KWIN100782730S KWIN100782730XL KWIN100782730XS KWIN100782930L KWIN100782930M KWIN100782930S KWIN100782930XS KWIN1007829302XL KWIN1007829303XL KWIN1007829304XL KWIN100782930XL KWIN1007829402XL KWIN1007829403XL KWIN1007829404XL KWIN100782940L KWIN100782940M KWIN100782940S KWIN100782940XL KWIN100782940XS
Sweatshirt, Match
Werkkleding
Durable sweatshirt made of 65% polyester and 35% cotton.
  • Double-dyed yarn and specially treated material against pilling and shrinkage
  • Shoulder trim
  • Stitching around upper arms and neck
  • Chest pocket
Weight: 300 g/m²