Kansas Icon Latzhose 2-873
Leverancier: Kwintet
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Kansas®
113-3020EA 93.4 EUR
113-3020 113-3021 113-3022 113-3027 113-3148 113-3149 113-3028 113-3029 113-3023 113-3024 113-3145 113-3146 113-3025 113-3147 113-3026 113-2979 113-2978 113-2977 113-3030 113-3151 113-3031 113-3152 113-3032 113-3153 113-3154 113-3033 113-3150 113-3038 113-2982 113-2981 113-3039 113-2980 113-3034 113-3155 113-3035 113-3156 113-2984 113-3036 113-2983 113-3037 113-3119 113-3080 113-3085 113-3086 113-3087 113-3000 113-3088 113-3121 113-3081 113-3082 113-3083 113-3084 113-3126 113-3005 113-3127 113-3128 113-3129 113-3089 113-3001 113-3122 113-3002 113-3123 113-3124 113-3003 113-3004 113-3125 113-3130 113-3131 113-3132 113-3019 113-3133 113-3134 113-3063 113-3064 113-3065 113-3066 113-3061 113-3062 113-3105 113-3106 113-3107 113-3067 113-3189 113-3068 113-3069 113-3108 113-3109 113-3190 113-3074 113-3075 113-3076 113-3110 113-3077 113-3191 113-3070 113-3071 113-3072 113-3073 113-3115 113-3116 113-3117 113-3118 113-3078 113-3111 113-3112 113-3079 113-3113 113-3114 113-3041 113-3042 113-3043 113-3044 113-3040 113-3045 113-2997 113-3046 113-2996 113-2995 113-3047 113-2999 113-2998 113-3173 113-3174 113-3175 113-3176 113-3170 113-3171 113-3172 113-3177
Kansas Icon Latzhose 2-873
Werkkleding
Fristads Kansas ICON bib trousers workwear for crafts and trades.
  • Inside breast pocket
  • Mobile phone pocket with velcro/pen pocket and hammer loop
  • Cordura® reinforced double ruler pocket
  • Two side pockets and two back pockets with creases - one with flap
  • Two-tone and contrast embroidery
  • Diagonally cut back section
  • Bib pocket with flap and leg pocket
Lateral access opening to the inner pants / diagonally cut knees. Reflective stripes under knee pockets and over leg closures. In the seams integrated knee pockets for height-adjustable knee pads with elastic carrier and adjustable waistband.
Material: 65% polyester, 35% cotton
Weight: 295 g/m²