Polo shirt, Match
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1007802102XL 1007802103XL 100780210L 100780210M 100780210S 100780210XL 100780210XS 1007803312XL 1007803313XL 1007803314XL 100780331L 100780331M 100780331S 100780331XL 100780331XS 1007805302XL 1007805303XL 1007805304XL 100780530L 100780530M 100780530S 100780530XL 100780530XS 1007805402XL 1007805403XL 1007805404XL 100780540L 100780540M 100780540S 100780540XL 100780540XS 1007807302XL 1007807303XL 1007807304XL 100780730L 100780730M 100780730S 100780730XL 100780730XS 1007809002XL 1007809003XL 1007809004XL 100780900L 100780900M 100780900S 100780900XL 100780900XS 1007809402XL 1007809403XL 1007809404XL 100780940L 100780940M 100780940S 100780940XL 100780940XS
KWIN1007802102XLEA 19.6 EUR
KWIN1007802102XL KWIN1007802103XL KWIN100780210L KWIN100780210M KWIN100780210S KWIN100780210XL KWIN100780210XS KWIN1007803312XL KWIN1007803313XL KWIN1007803314XL KWIN100780331L KWIN100780331M KWIN100780331S KWIN100780331XL KWIN100780331XS KWIN1007805302XL KWIN1007805303XL KWIN1007805304XL KWIN100780530L KWIN100780530M KWIN100780530S KWIN100780530XL KWIN100780530XS KWIN1007805402XL KWIN1007805403XL KWIN1007805404XL KWIN100780540L KWIN100780540M KWIN100780540S KWIN100780540XL KWIN100780540XS KWIN1007807302XL KWIN1007807303XL KWIN1007807304XL KWIN100780730L KWIN100780730M KWIN100780730S KWIN100780730XL KWIN100780730XS KWIN1007809002XL KWIN1007809003XL KWIN1007809004XL KWIN100780900L KWIN100780900M KWIN100780900S KWIN100780900XL KWIN100780900XS KWIN1007809402XL KWIN1007809403XL KWIN1007809404XL KWIN100780940L KWIN100780940M KWIN100780940S KWIN100780940XL KWIN100780940XS
Polo shirt, Match
Werkkleding
Basic polo shirt made of 65% polyester and 35% cotton. Available in many different colours.
  • Withstands many wash cycles
  • Button at neck
  • Reinforced shoulders
  • Chest pocket
Weight: 240 g/m²