3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: CHAPS
DG049 DG096
DG049EA 132 EUR
DG049 DG096
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formule: C₃₂H₅₈N₂O₇S
Smeltpunt: 116 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00081083
CAS nummer: 75621-03-3
Merck Index: 13,02052