N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3-(N,N-Dimethylmyristylammonio)propanesulphonate, Tetradecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner Salt

35867.01 35867.02
SERA35867.01EA 0 EUR
SERA35867.01 SERA35867.02
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (sulphobetaine-14, SB-14)
Formule: C₁₉H₄₁NO₃S
MW: 363.5988 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00036910
CAS nummer: 14933-09-6
EINECS: 239-003-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR