AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
328110500 328110010
ACRO328110500EA 55.7 EUR
ACRO328110500 ACRO328110010
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 98%
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formule: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Smeltpunt: 175…177 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00132962
CAS nummer: 30827-99-7
UN: 3261
ADR: 8 II
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %