AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
H26473.MD
H26473.03
H26473.MDEA
208
EUR
H26473.MD
H26473.03
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride) 97%
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Purity: 97%
CAS No.: 30827-99-7
Molecular Formula: C8H10FNO2S.HCl
Formula Weight: 239.69
Storage Temperature: -30°C to -10°C
Melting Point: ca 184° dec.
Beilstein Reference: 7604627
UN No.: 3261
MDL No.: MFCD00132962
Formule:
C₈H₁₁ClFNO₂S MW: 239.69 g/mol Smeltpunt: ∼ 184 °C (ontbinding) Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD00132962 CAS nummer: 30827-99-7 UN: 3261 ADR: 8 II |