AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
A154.1 A154.2
ROTHA154.1EA 84.9 EUR
ROTHA154.1 ROTHA154.2
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Formule: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00132962
CAS nummer: 30827-99-7
UN: 3261
ADR: 8 II