Guanosine

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

24270.02 24270.03
SERA24270.02EA 0 EUR
SERA24270.02 SERA24270.03
Guanosine
Guanosine
Formule: C₁₀H₁₃N₅O₅
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010182
CAS nummer: 118-00-3
EINECS: 204-227-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR