Disinfectant wipes, Meliseptol® HBV Tissues
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18706 18707
148-4103EA 17.7 EUR
148-4103 148-4105
Disinfectant wipes, Meliseptol® HBV Tissues
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Desinfectiedoekjes
These ready-to-use disinfectant wipes for small alcohol resistant surfaces are saturated with fast-acting Meliseptol® rapid (based on n-propanol and quaternary ammonium compound).
  • Active against bacteria, mycobacteria (M. terrae), yeasts and viruses (HBV / HIV / HCV), Adeno-, Noro-, Polio-, Rota- and Vaccinia virus
  • Aldehyde- and amine-free
  • Available in reclosable dispenser box and as an economic refill pack
  • Active in 1 minute
Use biocides safely. Always read the label and product information before use.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904