Magnetic stirring bars, plain, PTFE
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001.212 001.215.6 001.230 001.225.8 001.250.7 001.235.7 001.250 001.260
COWI001.212EA 1 EUR
COWI001.212 COWI001.215.6 COWI001.230 COWI001.225.8 COWI001.250.7 COWI001.235.7 COWI001.250 COWI001.260
Magnetic stirring bars, plain, PTFE
Roerders Magnetische roerstaven
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.