Pump tubing, PharMed®
Leverancier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
525-20012-00 525-20013-00 525-20015-00 525-20016-00 525-20022-00 525-20025-00 525-20026-00 525-20027-00 525-20028-00 525-22000-00 525-23000-00 525-24000-00 525-26000-00 525-29000-00
228-5201EA 25.4 EUR
228-5201 228-5202 228-5204 228-5205 228-0497 228-0500 228-0501 HEID525-20027-00 HEID525-20028-00 224-1476 224-1472 224-1473 224-1475 HEID525-29000-00
Pump tubing, PharMed®
Slangen Peristaltische pompslangen
PharMed®, thermoplastic elastomer on a polyproylene basis with plasticizers; excellent tensile strength, opaque.
  • High fatigue strength under repeated reversed bending stresses
  • Non-toxic, biocompatible
  • Very low gas permeability
  • Well suited to acids and bases
Ideal for medical, lab and research uses.
Certificaten: Complies with the standards: FDA (21 CFR 177.2600), USP 23 Class VI, GLP conform.