Tubing connectors, L-shaped, Bayonet type
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
016.1005.2 016.1002.2 016.1002.5.2 016.1003.2 016.1003.7.2 016.1004.9.2 016.1004.2
COWI016.1005.2EA 16 EUR
COWI016.1005.2 COWI016.1002.2 COWI016.1002.5.2 COWI016.1003.2 COWI016.1003.7.2 COWI016.1004.9.2 COWI016.1004.2
Tubing connectors, L-shaped, Bayonet type
Fittingen en connectoren
L-shaped right angle connectors, made of PTFE, for tubings.
  • Very good chemical resistance
  • Very good temperature resistance