Tubing connectors, 4-way
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
016.1115.2 016.1114.9.2 016.1114.2 016.1113.7.2 016.1113.2 016.1112.5.2 016.1112.2
COWI016.1115.2EA 19.3 EUR
COWI016.1115.2 COWI016.1114.9.2 COWI016.1114.2 COWI016.1113.7.2 COWI016.1113.2 COWI016.1112.5.2 COWI016.1112.2
Tubing connectors, 4-way
Fittingen en connectoren
4-way connectors, made of PTFE, bayonet type for tubings.
  • Very good chemical resistance
  • Very good temperature resistance