Clips for spherical ground glass joints
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.133.013 0.133.019 0.133.035 0.133.029 0.133.040 0.133.051 0.133.064
WITG0.133.013EA 12.4 EUR
WITG0.133.013 WITG0.133.019 WITG0.133.035 WITG0.133.029 WITG0.133.040 WITG0.133.051 WITG0.133.064
Clips for spherical ground glass joints
Klemmen NS-verbindingsklemmen
Very strong clips for spherical ground glass joints, made of stainless steel. With screw.