Adapters, reduction, Preciso
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.500.005 0.500.020
WITG0.500.005EA 8.9 EUR
WITG0.500.005 WITG0.500.020
Adapters, reduction, Preciso
Adapters Adapters met NS-verbindingen
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Certificaten: According to DIN 12257.