Adapters, Combifix®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5206634 5206642
720-2583EA 351 EUR
720-2583 BRAU5206642
Adapters, Combifix®
Fittingen en connectoren
These adapters are used for connecting fittings.
Suitable for interconnecting male and female fittings with 6% (Luer) and 10% (Rekord) taper fittings.