Stoppers, flat type, PTFE
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
010.134 010.129 010.124 010.119 010.114 010.110
COWI010.134EA 22.2 EUR
COWI010.134 COWI010.129 COWI010.124 COWI010.119 COWI010.114 COWI010.110
Stoppers, flat type, PTFE
Stoppen PE/LDPE/HDPE stoppen
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure