Short adapters for conical ground joints
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.540.006 0.540.005 0.540.001 0.540.007 0.540.008 0.540.002 0.540.004 0.540.003
WITG0.540.006EA 39.8 EUR
WITG0.540.006 WITG0.540.005 WITG0.540.001 WITG0.540.007 WITG0.540.008 WITG0.540.002 WITG0.540.004 WITG0.540.003
Short adapters for conical ground joints
Adapters Adapters met NS-verbindingen
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Short adapters, socket in cone.
Order Now
Specificaties
Environmentally Preferable