Rotary evaporating flasks, with standard ground joints
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.658.050 0.658.010 0.658.025
WITG0.658.050EA 9.9 EUR
WITG0.658.050 WITG0.658.010 WITG0.658.025
Rotary evaporating flasks, with standard ground joints
Kolven Kolven voor rotatieverdampers
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Pear-shaped with round bottom, centered.