Flasks, round bottom, with three necks, side necks angled (20°)
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0.736.002 0.730.001 0.732.001 0.732.500 0.725.025 0.725.050
WITG0.736.002EA 95.5 EUR
WITG0.736.002 WITG0.730.001 WITG0.732.001 WITG0.732.500 WITG0.725.025 WITG0.725.050
Flasks, round bottom, with three necks, side necks angled (20°)
Kolven Rondbodemkolven
Borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Certificaten: DIN 12394