Capillary columns, Elite-1301 for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9316210 N9316214 N9316215 N9316217 N9316219 N9316221 N9316223 N9316227
PERCN9316210EA 567 EUR
PERCN9316210 PERCN9316214 PERCN9316215 PERCN9316217 PERCN9316219 PERCN9316221 PERCN9316223 PERCN9316227
Capillary columns, Elite-1301 for autosystem
Kolommen voor chromatografie
Elite-1301 (6% Cyanopropylphenyl) Methylpolysiloxane series capillary columns are fused silica capillary columns ideal for the analysis of nonpolar to polar compounds and acid to basic species.